Learn More
Biolegend Alexa Fluor 647 anti-Histone H3 (C-terminus)
Supplier:  Biolegend 819408
Encompass_Preferred
Alexa Fluor 647 anti-Histone H3 (C-terminus) [1B1-B2]; Isotype: Mouse IgG3; Reactivity: Human, Mouse, Rat, Drosophila; Apps: IHC-P; Size: 100 ug
Catalog No. 50-207-4148