Learn More
Biolegend Alexa Fluor 647 anti-human CD14
Supplier:  Biolegend 325612
Encompass_Preferred
Alexa Fluor 647 anti-human CD14 [HCD14]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ICC; Size: 100 tests
Catalog No. 50-167-297