Learn More
Biolegend Alexa Fluor 647 anti-human/rat CD51 (Integrin αvβ5)
Supplier:  Biolegend 920006
Encompass_Preferred
Alexa Fluor 647 anti-human/rat CD51 (Integrin αvβ5) [P1F6]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rat, Cross-Reactivity: Hamster; Apps: FC; Size: 100 tests
Catalog No. 50-207-5230