Learn More
Biolegend Alexa Fluor 647 anti-mouse Blimp-1
Supplier:  Biolegend 150004
Encompass_Preferred
Alexa Fluor 647 anti-mouse Blimp-1 [5E7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, WB; Size: 100 ug
Catalog No. 50-207-1907