Learn More
Biolegend Alexa Fluor 647 anti-mouse CD3ε
Supplier:  Biolegend 100322
Encompass_Preferred
Alexa Fluor 647 anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F; Size: 100 ug
Catalog No. 50-162-398