Learn More
Biolegend Alexa Fluor 647 anti-mouse/human Mac-2 (Galectin-3)
Supplier:  Biolegend 125408
Encompass_Preferred
Alexa Fluor 647 anti-mouse/human Mac-2 (Galectin-3) [M3/38]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human; Apps: FC, IHC-P; Size: 100 ug
Catalog No. 50-164-252