Learn More
Apexbio Technology LLC A66, 10mg. Cas: 1166227-08-2 MFCD: MFCD22378485. P110α inhibitor
SDP
Supplier:  Apexbio Technology LLC A835410
Encompass_Preferred
A66 is a potent and selective p110 inhibitor with IC50 of 32 nM.P110 is the catalytic subunit of a class I Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic that is composed of a 85 kDa regulatory subunit and a 110 kDa catalytic subunit. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5125