Learn More
Medchemexpress LLC HY-13261 5mg Medchemexpress, A66 CAS:1166227-08-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132615MG
Encompass_Preferred
Medchemexpress, HY-13261 5mg A66 CAS:1166227-08-2 A66 is a highly specific and selective p110α inhibitor with an IC50 of 32 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1319