Learn More
ABclonal Technology NEK2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A68111000UL
Encompass_Preferred
This gene encodes a serine/threonine-protein kinase that is involved in mitotic regulation. This protein is localized to the centrosome, and undetectable during G1 phase, but accumulates progressively throughout the S phase, reaching maximal levels in late G2 phase. Alternatively spliced transcript variants encoding different isoforms with distinct C-termini have been noted for this gene.
Catalog No. 50-001-35516