Learn More
ABclonal Technology CDKN1C/p57 Kip2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A6843500UL
Encompass_Preferred
This gene is imprinted, with preferential expression of the maternal allele. The encoded protein is a tight-binding, strong inhibitor of several G1 cyclin/Cdk complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndorome, suggesting that this gene is a tumor suppressor candidate. Three transcript variants encoding two different isoforms have been found for this gene.
Catalog No. 50-001-37014