Learn More
Medchemexpress LLC A68930 hydrochloride | 130465-39-3 | MFCD05863727 | 98.7% | 307.77 | C16H18ClNO3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10343110MM 1ML
Encompass_Preferred
A68930 hydrochloride is a selective dopamine D1 receptor agonist used in research of D1 receptor pharmacology and respiratory models such as bronchiectasis. It is supplied as a solid or as a 10 mM solution in DMSO, with reported purity of approximately 98.7% and molecular weight 307.77.
  • Selective dopamine D1 receptor agonist
  • Available as 10 mM solution in DMSO or as solid samples
  • High reported purity (~98.7%) suitable for research use
  • Used in pharmacology and respiratory disease research
  • Molecular formula C16H18ClNO3; molecular weight 307.77
Catalog No. 50-002-46364