Learn More
Medchemexpress LLC A68930 hydrochloride | 130465-39-3 | MFCD05863727 | 98.7% | 307.77 g/mol | C16H18ClNO3 | 1 MG

Supplier: Medchemexpress LLC HY1034311MG
A68930 hydrochloride is a dopamine D1 receptor agonist supplied for research use. It elevates intracellular cAMP, increases CREB phosphorylation, and induces MUC5AC expression in airway epithelial cell models, making it useful in bronchiectasis and related respiratory research.
- Mechanism: dopamine D1 receptor agonist
- Applications: bronchiectasis and airway epithelial cell research
- Molecular formula: C16H18ClNO3
- Molecular weight: 307.77 g/mol
- Purity: 98.71%
- Physical form: white to yellow solid; solution option available
- Pack sizes: 1 MG and 5 MG solid; 10 mM 1 ML solution
- Storage: solid at 4°C sealed; solutions -80°C (6 months) or -20°C (1 month)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.