Learn More
Medchemexpress LLC A71623 CCK-A full agonist | 130408-77-4 | 99.3% | 840.96 | 1 MG

Supplier: Medchemexpress LLC HYP10961MG
A71623, a CCK-4-based peptide, is a potent and highly selective CCK-A full agonist. It shows high affinity for CCK-A receptors and modulates food intake and motor performance in various animal models. This peptide is suitable for research in neuroscience and metabolic studies.
- Potent and highly selective CCK-A full agonist
- IC50 of 3.7 nM in guinea pig pancreas (CCK-A)
- Reduces food intake and suppresses liquid diet intake in rats
- Dampens Purkinje neuron pathology in SCA mice models
- Improves motor performance in SCA2 mice
- Soluble in DMSO (50 mg/mL)
- Chemical formula: C44H56N8O9
- Molecular weight: 840.96
- Appearance: White to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.