Learn More
ABclonal Technology MOV10 Rabbit mAb
SDP
Supplier:  ABclonal Technology A7227500UL
Encompass_Preferred
Enables 5-3 RNA helicase activity and RNA binding activity. Involved in defense response to virus, negative regulation of transposition, RNA-mediated, and posttranscriptional regulation of gene expression. Located in P-body and cytosol. Implicated in hypertension.
Catalog No. 50-001-35599