Learn More
ABclonal Technology LysRS/KARS Rabbit mAb
SDP
Supplier:  ABclonal Technology A8648500UL
Encompass_Preferred
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-34774