Learn More
ABclonal Technology eIF2B epsilon (EIF2B5) Rabbit mAb
SDP
Supplier:  ABclonal Technology A86701000UL
Encompass_Preferred
This gene encodes one of five subunits of eukaryotic translation initiation factor 2B (EIF2B), a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.
Catalog No. 50-001-34893