Learn More
ABclonal Technology NDUFS4 Rabbit mAb
SDP
Supplier:  ABclonal Technology A8691500UL
Encompass_Preferred
This gene encodes an nuclear-encoded accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (complex I, or NADHubiquinone oxidoreductase). Complex I removes electrons from NADH and passes them to the electron acceptor ubiquinone. Mutations in this gene can cause mitochondrial complex I deficiencies such as Leigh syndrome. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-34894