Learn More
ABclonal Technology OPRD1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A86941000UL
Encompass_Preferred
Enables G protein-coupled enkephalin receptor activity. Involved in several processes, including G protein-coupled opioid receptor signaling pathway, cellular response to hypoxia, and positive regulation of peptidyl-serine phosphorylation. Is intrinsic component of plasma membrane.
Catalog No. 50-001-34638