Learn More
ABclonal Technology FXR1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A86971000UL
Encompass_Preferred
The protein encoded by this gene is an RNA binding protein that interacts with the functionally-similar proteins FMR1 and FXR2. These proteins shuttle between the nucleus and cytoplasm and associate with polyribosomes, predominantly with the 60S ribosomal subunit. Three transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-34928