Learn More
ABclonal Technology FKBP38/FKBP8 Rabbit mAb
SDP
Supplier:  ABclonal Technology A87011000UL
Encompass_Preferred
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function.
Catalog No. 50-001-34598