Learn More
ABclonal Technology Calpain 1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A87101000UL
Encompass_Preferred
The calpains, calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene.
Catalog No. 50-001-34742