Learn More
ABclonal Technology MYL9 Rabbit mAb
SDP
Supplier:  ABclonal Technology A8738500UL
Encompass_Preferred
Myosin, a structural component of muscle, consists of two heavy chains and four light chains. The protein encoded by this gene is a myosin light chain that may regulate muscle contraction by modulating the ATPase activity of myosin heads. The encoded protein binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-36823