Learn More
ABclonal Technology RanGAP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A87581000UL
Encompass_Preferred
This gene encodes a protein that associates with the nuclear pore complex and participates in the regulation of nuclear transport. The encoded protein interacts with Ras-related nuclear protein 1 (RAN) and regulates guanosine triphosphate (GTP)-binding and exchange. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-36843