Learn More
ABclonal Technology [KO Validated] HPRT1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A8783500UL
Encompass_Preferred
The protein encoded by this gene is a transferase, which catalyzes conversion of hypoxanthine to inosine monophosphate and guanine to guanosine monophosphate via transfer of the 5-phosphoribosyl group from 5-phosphoribosyl 1-pyrophosphate. This enzyme plays a central role in the generation of purine nucleotides through the purine salvage pathway. Mutations in this gene result in Lesch-Nyhan syndrome or gout.
Catalog No. 50-001-36880