Learn More
ABclonal Technology BUB3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A8831500UL
Encompass_Preferred
This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Catalog No. 50-001-36838