Learn More
ABclonal Technology Arp2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A88761000UL
Encompass_Preferred
The specific function of this gene has not yet been determined, however, the protein it encodes is known to be a major constituent of the ARP2/3 complex. This complex is located at the cell surface and is essential to cell shape and motility through lamellipodial actin assembly and protrusion. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-36840