Learn More
ABclonal Technology APPL1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A89321000UL
Encompass_Preferred
The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signalling and insulin signalling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
Catalog No. 50-001-34618