Learn More
ABclonal Technology Pyruvate carboxylase (PC) Rabbit mAb
SDP
Supplier:  ABclonal Technology A89801000UL
Encompass_Preferred
This gene encodes pyruvate carboxylase, which requires biotin and ATP to catalyse the carboxylation of pyruvate to oxaloacetate. The active enzyme is a homotetramer arranged in a tetrahedron which is located exclusively in the mitochondrial matrix. Pyruvate carboxylase is involved in gluconeogenesis, lipogenesis, insulin secretion and synthesis of the neurotransmitter glutamate. Mutations in this gene have been associated with pyruvate carboxylase deficiency. Alternatively spliced transcript variants with different 5 UTRs, but encoding the same protein, have been found for this gene.
Catalog No. 50-001-37143