Learn More
ABclonal Technology CXCR7 Rabbit mAb
SDP
Supplier:  ABclonal Technology A89911000UL
Encompass_Preferred
This gene encodes a member of the G-protein coupled receptor family. Although this protein was earlier thought to be a receptor for vasoactive intestinal peptide (VIP), it is now considered to be an orphan receptor, in that its endogenous ligand has not been identified. The protein is also a coreceptor for human immunodeficiency viruses (HIV). Translocations involving this gene and HMGA2 on chromosome 12 have been observed in lipomas.
Catalog No. 50-001-35610