Learn More
ABclonal Technology PELP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A90261000UL
Encompass_Preferred
This gene encodes a transcription factor which coactivates transcription of estrogen receptor responsive genes and corepresses genes activated by other hormone receptors or sequence-specific transcription factors. Expression of this gene is regulated by both members of the estrogen receptor family. This gene may be involved in the progression of several types of cancer. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-34581