Learn More
ABclonal Technology NUMB Rabbit mAb
SDP
Supplier:  ABclonal Technology A90391000UL
Encompass_Preferred
The protein encoded by this gene plays a role in the determination of cell fates during development. The encoded protein, whose degradation is induced in a proteasome-dependent manner by MDM2, is a membrane-bound protein that has been shown to associate with EPS15, LNX1, and NOTCH1. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-34670