Learn More
ABclonal Technology PROX1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A90471000UL
Encompass_Preferred
The protein encoded by this gene is a member of the homeobox transcription factor family. Members of this family contain a homeobox domain that consists of a 60-amino acid helix-turn-helix structure that binds DNA and RNA. The protein encoded by this gene is conserved across vertebrates and may play an essential role during development. Altered levels of this protein have been reported in cancers of different organs, such as colon, brain, blood, breast, pancreas, liver and esophagus. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-35543