Learn More
ABclonal Technology PTBP2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A91241000UL
Encompass_Preferred
The protein encoded by this gene binds to intronic polypyrimidine clusters in pre-mRNA molecules and is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein (PTB) but most of its isoforms are expressed primarily in the brain. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-36923