Learn More
ABclonal Technology NFIB / NF1B2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9125500UL
Encompass_Preferred
Enables DNA-binding transcription activator activity, RNA polymerase II-specific, RNA polymerase II cis-regulatory region sequence-specific DNA binding activity, and transcription regulator inhibitor activity. Involved in brain development, negative regulation of DNA binding activity, and regulation of transcription by RNA polymerase II. Located in fibrillar center and nucleoplasm.
Catalog No. 50-001-36830