Learn More
ABclonal Technology KIF2C/MCAK Rabbit mAb
SDP
Supplier:  ABclonal Technology A91401000UL
Encompass_Preferred
This gene encodes a kinesin-like protein that functions as a microtubule-dependent molecular motor. The encoded protein can depolymerize microtubules at the plus end, thereby promoting mitotic chromosome segregation. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-36960