Learn More
ABclonal Technology eIF3B Rabbit mAb
SDP
Supplier:  ABclonal Technology A9143500UL
Encompass_Preferred
Enables RNA binding activity. Contributes to translation initiation factor activity. Involved in several processes, including IRES-dependent viral translational initiation, translational initiation, and viral translational termination-reinitiation. Located in extracellular exosome. Part of eukaryotic translation initiation factor 3 complex.
Catalog No. 50-001-36938