Learn More
ABclonal Technology Carbonic Anhydrase 2 (CA2) Rabbit mAb
SDP
Supplier:  ABclonal Technology A91481000UL
Encompass_Preferred
The protein encoded by this gene is one of several isozymes of carbonic anhydrase, which catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-35606