Learn More
ABclonal Technology GRK3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A91631000UL
Encompass_Preferred
The beta-adrenergic receptor kinase specifically phosphorylates the agonist-occupied form of the beta-adrenergic and related G protein-coupled receptors. Overall, the beta adrenergic receptor kinase 2 has 85% amino acid similarity with beta adrenergic receptor kinase 1, with the protein kinase catalytic domain having 95% similarity. These data suggest the existence of a family of receptor kinases which may serve broadly to regulate receptor function.
Catalog No. 50-001-35469