Learn More
ABclonal Technology Profilin1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A91881000UL
Encompass_Preferred
This gene encodes a member of the profilin family of small actin-binding proteins. The encoded protein plays an important role in actin dynamics by regulating actin polymerization in response to extracellular signals. Deletion of this gene is associated with Miller-Dieker syndrome, and the encoded protein may also play a role in Huntington disease. Multiple pseudogenes of this gene are located on chromosome 1.
Catalog No. 50-001-35640