Learn More
ABclonal Technology [KO Validated] IMPDH2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A92081000UL
Encompass_Preferred
This gene encodes the rate-limiting enzyme in the de novo guanine nucleotide biosynthesis. It is thus involved in maintaining cellular guanine deoxy- and ribonucleotide pools needed for DNA and RNA synthesis. The encoded protein catalyzes the NAD-dependent oxidation of inosine-5-monophosphate into xanthine-5-monophosphate, which is then converted into guanosine-5-monophosphate. This gene is up-regulated in some neoplasms, suggesting it may play a role in malignant transformation.
Catalog No. 50-001-36827