Learn More
ABclonal Technology PIM2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A92301000UL
Encompass_Preferred
This gene encodes a protooncogene that acts as a serine/threonine protein kinase. Studies determined the encoded protein functions to prevent apoptosis and to promote cell survival.
Catalog No. 50-001-35614