Learn More
ABclonal Technology Podoplanin Rabbit mAb
SDP
Supplier:  ABclonal Technology A92421000UL
Encompass_Preferred
This gene encodes a type-I integral membrane glycoprotein with diverse distribution in human tissues. The physiological function of this protein may be related to its mucin-type character. The homologous protein in other species has been described as a differentiation antigen and influenza-virus receptor. The specific function of this protein has not been determined but it has been proposed as a marker of lung injury. Alternatively spliced transcript variants encoding different isoforms have been identified.
Catalog No. 50-001-36806