Learn More
ABclonal Technology UBA1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A92541000UL
Encompass_Preferred
The protein encoded by this gene catalyzes the first step in ubiquitin conjugation to mark cellular proteins for degradation. This gene complements an X-linked mouse temperature-sensitive defect in DNA synthesis, and thus may function in DNA repair. It is part of a gene cluster on chromosome Xp11.23. Alternatively spliced transcript variants that encode the same protein have been described.
Catalog No. 50-001-35616