Learn More
ABclonal Technology USP22 Rabbit mAb
SDP
Supplier:  ABclonal Technology A9261500UL
Encompass_Preferred
Enables enzyme binding activity, nuclear receptor coactivator activity, and thiol-dependent deubiquitinase. Contributes to H4 histone acetyltransferase activity. Involved in positive regulation of mitotic cell cycle, positive regulation of transcription, DNA-templated, and protein modification by small protein conjugation or removal. Part of SAGA complex.
Catalog No. 50-001-37258