Learn More
ABclonal Technology CRALBP Rabbit mAb
SDP
Supplier:  ABclonal Technology A92651000UL
Encompass_Preferred
The protein encoded by this gene is a 36-kD water-soluble protein which carries 11-cis-retinaldehyde or 11-cis-retinal as physiologic ligands. It may be a functional component of the visual cycle. Mutations of this gene have been associated with severe rod-cone dystrophy, Bothnia dystrophy (nonsyndromic autosomal recessive retinitis pigmentosa) and retinitis punctata albescens.
Catalog No. 50-001-34801