Learn More
ABclonal Technology KDM4A Rabbit mAb
SDP
Supplier:  ABclonal Technology A92671000UL
Encompass_Preferred
This gene is a member of the Jumonji domain 2 (JMJD2) family and encodes a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This nuclear protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
Catalog No. 50-001-34647