Learn More
ABclonal Technology Nuclear Matrix Protein p84 (THOC1) Rabbit mAb
SDP
Supplier:  ABclonal Technology A9269500UL
Encompass_Preferred
Predicted to enable DNA binding activity and RNA binding activity. Involved in several processes, including negative regulation of DNA damage checkpoint, regulation of nucleobase-containing compound metabolic process, and viral mRNA export from host cell nucleus. Located in cytoplasm and nuclear speck. Part of THO complex part of transcription export complex. Colocalizes with chromosome, telomeric region.
Catalog No. 50-001-37129