Learn More
ABclonal Technology FOXO3A Rabbit mAb
SDP
Supplier:  ABclonal Technology A9270500UL
Encompass_Preferred
This gene belongs to the forkhead family of transcription factors which are characterized by a distinct forkhead domain. This gene likely functions as a trigger for apoptosis through expression of genes necessary for cell death. Translocation of this gene with the MLL gene is associated with secondary acute leukemia. Alternatively spliced transcript variants encoding the same protein have been observed.
Catalog No. 50-001-36436