Learn More
ABclonal Technology DNA Ligase I Rabbit mAb
SDP
Supplier:  ABclonal Technology A9301500UL
Encompass_Preferred
This gene encodes a member of the ATP-dependent DNA ligase protein family. The encoded protein functions in DNA replication, recombination, and the base excision repair process. Mutations in this gene that lead to DNA ligase I deficiency result in immunodeficiency and increased sensitivity to DNA-damaging agents. Disruption of this gene may also be associated with a variety of cancers. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-36141