Learn More
ABclonal Technology STXBP1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A93091000UL
Encompass_Preferred
This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
Catalog No. 50-001-35567